Merck
MAFG-driven astrocytes promote CNS inflammation.
Nature (2020-02-14)
Michael A Wheeler, Iain C Clark, Emily C Tjon, Zhaorong Li, Stephanie E J Zandee, Charles P Couturier, Brianna R Watson, Giulia Scalisi, Sarah Alkwai, Veit Rothhammer, Assaf Rotem, John A Heyman, Shravan Thaploo, Liliana M Sanmarco, Jiannis Ragoussis, David A Weitz, Kevin Petrecca, Jeffrey R Moffitt, Burkhard Becher, Jack P Antel, Alexandre Prat, Francisco J Quintana
ABSTRACT
Multiple sclerosis is a chronic inflammatory disease of the CNS1. Astrocytes contribute to the pathogenesis of multiple sclerosis2, but little is known about the heterogeneity of astrocytes and its regulation. Here we report the analysis of astrocytes in multiple sclerosis and its preclinical model experimental autoimmune encephalomyelitis (EAE) by single-cell RNA sequencing in combination with cell-specific Ribotag RNA profiling, assay for transposase-accessible chromatin with sequencing (ATAC-seq), chromatin immunoprecipitation with sequencing (ChIP-seq), genome-wide analysis of DNA methylation and in vivo CRISPR-Cas9-based genetic perturbations. We identified astrocytes in EAE and multiple sclerosis that were characterized by decreased expression of NRF2 and increased expression of MAFG, which cooperates with MAT2α to promote DNA methylation and represses antioxidant and anti-inflammatory transcriptional programs. Granulocyte-macrophage colony-stimulating factor (GM-CSF) signalling in astrocytes drives the expression of MAFG and MAT2α and pro-inflammatory transcriptional modules, contributing to CNS pathology in EAE and, potentially, multiple sclerosis. Our results identify candidate therapeutic targets in multiple sclerosis.
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
1H,1H,2H,2H-Perfluoro-1-octanol, 97%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Millipore
Immobilon®-P PVDF Membrane, 10 sheets, 15 cm x 15 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-MAFG antibody produced in rabbit
Sigma-Aldrich
Ionomycin from Streptomyces conglobatus, ≥98% (HPLC)
Hamilton® syringe, 700 series, removable needle, 702RN, volume 25 μL, needle size 22s ga (bevel tip), needle L 51 mm (2 in.)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Trizma® hydrochloride solution, 1 M, BioReagent, for molecular biology
Sigma-Aldrich
Earle′s Balanced Salt Solution 10x, Without sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
DAPI, for nucleic acid staining