Merck
  • Intracellular directed evolution of proteins from combinatorial libraries based on conditional phage replication.
Intracellular directed evolution of proteins from combinatorial libraries based on conditional phage replication.
Nature protocols (2017-08-11)
Andreas K Brödel, Alfonso Jaramillo, Mark Isalan
ABSTRACT
Directed evolution is a powerful tool to improve the characteristics of biomolecules. Here we present a protocol for the intracellular evolution of proteins with distinct differences and advantages in comparison with established techniques. These include the ability to select for a particular function from a library of protein variants inside cells, minimizing undesired coevolution and propagation of nonfunctional library members, as well as allowing positive and negative selection logics using basally active promoters. A typical evolution experiment comprises the following stages: (i) preparation of a combinatorial M13 phagemid (PM) library expressing variants of the gene of interest (GOI) and preparation of the Escherichia coli host cells; (ii) multiple rounds of an intracellular selection process toward a desired activity; and (iii) the characterization of the evolved target proteins. The system has been developed for the selection of new orthogonal transcription factors (TFs) but is capable of evolving any gene-or gene circuit function-that can be linked to conditional M13 phage replication. Here we demonstrate our approach using as an example the directed evolution of the bacteriophage λ cI TF against two synthetic bidirectional promoters. The evolved TF variants enable simultaneous activation and repression against their engineered promoters and do not cross-react with the wild-type promoter, thus ensuring orthogonality. This protocol requires no special equipment, allowing synthetic biologists and general users to evolve improved biomolecules within ∼7 weeks.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Carbenicillin disodium salt, 89.0-100.5% anhydrous basis
Sigma-Aldrich
KOD Hot Start DNA Polymerase, High fidelity DNA polymerase designed for accurate PCR amplification of long strand and GC- rich DNA templates for cloning and cDNA amplification applications.
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%