Merck
  • Class IIa Histone Deacetylases Drive Toll-like Receptor-Inducible Glycolysis and Macrophage Inflammatory Responses via Pyruvate Kinase M2.
Class IIa Histone Deacetylases Drive Toll-like Receptor-Inducible Glycolysis and Macrophage Inflammatory Responses via Pyruvate Kinase M2.
Cell reports (2020-02-27)
Kaustav Das Gupta, Melanie R Shakespear, James E B Curson, Ambika M V Murthy, Abishek Iyer, Mark P Hodson, Divya Ramnath, Vikas A Tillu, Jessica B von Pein, Robert C Reid, Kathryn Tunny, Daniel M Hohenhaus, Shayli Varasteh Moradi, Gregory M Kelly, Takumi Kobayashi, Jennifer H Gunter, Alexander J Stevenson, Weijun Xu, Lin Luo, Alun Jones, Wayne A Johnston, Antje Blumenthal, Kirill Alexandrov, Brett M Collins, Jennifer L Stow, David P Fairlie, Matthew J Sweet
ABSTRACT
Histone deacetylases (HDACs) drive innate immune cell-mediated inflammation. Here we identify class IIa HDACs as key molecular links between Toll-like receptor (TLR)-inducible aerobic glycolysis and macrophage inflammatory responses. A proteomic screen identified the glycolytic enzyme pyruvate kinase M isoform 2 (Pkm2) as a partner of proinflammatory Hdac7 in murine macrophages. Myeloid-specific Hdac7 overexpression in transgenic mice amplifies lipopolysaccharide (LPS)-inducible lactate and promotes a glycolysis-associated inflammatory signature. Conversely, pharmacological or genetic targeting of Hdac7 and other class IIa HDACs attenuates LPS-inducible glycolysis and accompanying inflammatory responses in macrophages. We show that an Hdac7-Pkm2 complex acts as an immunometabolism signaling hub, whereby Pkm2 deacetylation at lysine 433 licenses its proinflammatory functions. Disrupting this complex suppresses inflammatory responses in vitro and in vivo. Class IIa HDACs are thus pivotal intermediates connecting TLR-inducible glycolysis to inflammation via Pkm2.
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by gel-filtration chromatography
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
PKM2 Activator II, DASA, The PKM2 Activator II, DASA modulates the biological activity of PKM2. This small molecule/inhibitor is primarily used for Biochemicals applications.