Merck
  • The Thalamus Regulates Retinoic Acid Signaling and Development of Parvalbumin Interneurons in Postnatal Mouse Prefrontal Cortex.
The Thalamus Regulates Retinoic Acid Signaling and Development of Parvalbumin Interneurons in Postnatal Mouse Prefrontal Cortex.
eNeuro (2019-03-15)
Rachel Larsen, Alatheia Proue, Earl Parker Scott, Matthew Christiansen, Yasushi Nakagawa
ABSTRACT
GABAergic inhibitory neurons in the prefrontal cortex (PFC) play crucial roles in higher cognitive functions. Despite the link between aberrant development of PFC interneurons and a number of psychiatric disorders, mechanisms underlying the development of these neurons are poorly understood. Here we show that the retinoic acid (RA)-degrading enzyme CYP26B1 (cytochrome P450 family 26, subfamily B, member 1) is transiently expressed in the mouse frontal cortex during postnatal development, and that medial ganglionic eminence (MGE)-derived interneurons, particularly in parvalbumin (PV)-expressing neurons, are the main cell type that has active RA signaling during this period. We found that frontal cortex-specific Cyp26b1 knock-out mice had an increased density of PV-expressing, but not somatostatin-expressing, interneurons in medial PFC, indicating a novel role of RA signaling in controlling PV neuron development. The initiation of Cyp26b1 expression in neonatal PFC coincides with the establishment of connections between the thalamus and the PFC. We found that these connections are required for the postnatal expression of Cyp26b1 in medial PFC. In addition to this region-specific role in postnatal PFC that regulates RA signaling and PV neuron development, the thalamocortical connectivity had an earlier role in controlling radial dispersion of MGE-derived interneurons throughout embryonic neocortex. In summary, our results suggest that the thalamus plays multiple, temporally separate roles in interneuron development in the PFC.
Sigma-Aldrich
Anti-Tbr1 Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®