Merck
  • Single-Cell Transcriptomics Uncovers Zonation of Function in the Mesenchyme during Liver Fibrosis.
Single-Cell Transcriptomics Uncovers Zonation of Function in the Mesenchyme during Liver Fibrosis.
Cell reports (2019-11-14)
Ross Dobie, John R Wilson-Kanamori, Beth E P Henderson, James R Smith, Kylie P Matchett, Jordan R Portman, Karolina Wallenborg, Simone Picelli, Anna Zagorska, Swetha V Pendem, Thomas E Hudson, Minnie M Wu, Grant R Budas, David G Breckenridge, Ewen M Harrison, Damian J Mole, Stephen J Wigmore, Prakash Ramachandran, Chris P Ponting, Sarah A Teichmann, John C Marioni, Neil C Henderson
ABSTRACT
Iterative liver injury results in progressive fibrosis disrupting hepatic architecture, regeneration potential, and liver function. Hepatic stellate cells (HSCs) are a major source of pathological matrix during fibrosis and are thought to be a functionally homogeneous population. Here, we use single-cell RNA sequencing to deconvolve the hepatic mesenchyme in healthy and fibrotic mouse liver, revealing spatial zonation of HSCs across the hepatic lobule. Furthermore, we show that HSCs partition into topographically diametric lobule regions, designated portal vein-associated HSCs (PaHSCs) and central vein-associated HSCs (CaHSCs). Importantly we uncover functional zonation, identifying CaHSCs as the dominant pathogenic collagen-producing cells in a mouse model of centrilobular fibrosis. Finally, we identify LPAR1 as a therapeutic target on collagen-producing CaHSCs, demonstrating that blockade of LPAR1 inhibits liver fibrosis in a rodent NASH model. Taken together, our work illustrates the power of single-cell transcriptomics to resolve the key collagen-producing cells driving liver fibrosis with high precision.
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Gey′s Balanced Salt Solution, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Chloramine-T hydrate, 95%
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Avanti
18:2 Lyso PA, Avanti Research - A Croda Brand
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid