Merck
  • A Steric Gating Mechanism Dictates the Substrate Specificity of a Rab-GEF.
A Steric Gating Mechanism Dictates the Substrate Specificity of a Rab-GEF.
Developmental cell (2018-12-12)
Laura L Thomas, Solveig A van der Vegt, J Christopher Fromme
ABSTRACT
Correct localization of Rab GTPases in cells is critical for proper function in membrane trafficking, yet the mechanisms that target Rabs to specific subcellular compartments remain controversial. Guanine nucleotide exchange factors (GEFs) activate and consequently stabilize Rab substrates on membranes, thus implicating GEFs as the primary determinants of Rab localization. A competing hypothesis is that the Rab C-terminal hypervariable domain (HVD) serves as a subcellular targeting signal. In this study, we present a unifying mechanism in which the HVD controls targeting of certain Rabs by mediating interaction with their GEFs. We demonstrate that the TRAPP complexes, two related GEFs that use the same catalytic site to activate distinct Rabs, distinguish between Ypt1 (Rab1) and Ypt31/32 (Rab11) via their divergent HVDs. Remarkably, we find that HVD length gates Rab access to the TRAPPII complex by constraining the distance between the nucleotide-binding domain and the membrane surface.
Sigma-Aldrich
N-Stearoyl-D-sphingosine, ≥98.0% (TLC)
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycerol, ≥97%
Sigma-Aldrich
Magnesium acetate solution, BioUltra, for molecular biology, ~1 M in H2O
Avanti
16:0 CDP DG, Avanti Research - A Croda Brand 870510P, powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)