Merck
Cell cycle-dependent activation of Ras.
Current biology : CB (1996-12-01)
S J Taylor, D Shalloway
ABSTRACT
Ras proteins play an essential role in the transduction of signals from a wide range of cell-surface receptors to the nucleus. These signals may promote cellular proliferation or differentiation, depending on the cell background. It is well established that Ras plays an important role in the transduction of mitogenic signals from activated growth-factor receptors, leading to cell-cycle entry. However, important questions remain as to whether Ras controls signalling events during cell-cycle progression and, if so, at which point in the cell-cycle it is activated. To address these questions we have developed a novel, functional assay for the detection of cellular activated Ras. Using this assay, we found that Ras was activated in HeLa cells, following release from mitosis, and in NIH 3T3 fibroblasts, following serum-stimulated cell-cycle entry. In each case, peak Ras activation occurred in mid-G1 phase. Ras activation in HeLa cells at mid-G1 phase was dependent on RNA and protein synthesis and was not associated with tyrosine phosphorylation of Shc proteins and their binding to Grb2. Significantly, activation of Ras and the extracellular-signal regulated (ERK) sub-group of mitogen-activated protein kinases were not temporally correlated during G1-phase progression. Activation of Ras during mid-G1 phase appears to differ in many respects from its rapid activation by growth factors, suggesting a novel mechanism of regulation that may be intrinsic to cell-cycle progression. Furthermore, the temporal dissociation between Ras and ERK activation suggests that Ras targets alternate effector pathways during G1-phase progression.
Sigma-Aldrich
Rac/cdc42 Assay Reagent (PAK1 PBD, agarose), 300 µg, Specifically binds to & precipitates Rac-GTP & cdc42-GTP from cell lysates. For use in Affinity Binding Assays.
Sigma-Aldrich
Ral Activation Assay Kit, Non-radioactive Ral Activation Assay Kit can be used to precipitate Ral-A-GTP from cell lysates & detection by a Ral-A specific antibody.
Sigma-Aldrich
Cdc42 Activation Assay Kit, Non-radioactive cdc42 Activation Assay Kit can be used to precipitate cdc42-GTP from cell lysates subjected to various treatments & detection by a cdc42 specific monoclonal antibody.
Sigma-Aldrich
Ras Assay Reagent (Raf-1 RBD, agarose)
Sigma-Aldrich
Ras Activation Assay Kit
Sigma-Aldrich
Rho Activation Assay Kit, PP1/PP2A Toolbox for the selective in vitro dephosphorylation of proteins.