Skip to Content
Merck
All Photos(1)

Key Documents

17-218

Sigma-Aldrich

Ras Activation Assay Kit

Synonym(s):

Ras Activity Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

manufacturer/tradename

Upstate®

Quality Level

technique(s)

activity assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... MRAS(22808)

General description

Muscle RAS oncogene homolog (MRAS) plays a key role in cardiovascular diseases. The MRAS gene codes for a member of the ras superfamily of GTP-binding proteins. MRAS is found in all tissues, particularly in the cardiovascular system. It is involved in signal transduction, including cell growth and differentiation.

Packaging

Kit capacity: 30 assays

Components

Ras Assay Reagent (Raf-1 RBD, agarose) (Cat.# 14-278)

Anti-Ras, clone RAS10 (Cat.# 05-516)

100X GTPγS, 10mM (Cat.# 20-176)

100X GDP, 100mM (Cat.# 20-177)

Mg2+ Lysis/Wash Buffer, 5X (Cat.# 20-168)

Quality

This kit precipitated Ras-GTP from HeLa cell lysates that had been loaded with GTPγS. The precipitated Ras-GTP was detected by western immunoblot using 1μg/ml anti-Ras, clone RAS10

Linkage

Replaces: SGT435

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AnhCo Nguyen et al.
Molecular and cellular biology, 22(9), 3035-3045 (2002-04-10)
While scaffold proteins are thought to be key components of signaling pathways, their exact function is unknown. By preassembling multiple components of signaling cascades, scaffolds are predicted to influence the efficiency and/or specificity of signaling events. Here we analyze a
X Shen et al.
The Biochemical journal, 352 Pt 3, 913-919 (2000-12-06)
Interleukin 1beta (IL-1beta) suppresses the IL-6-dependent induction of type II acute-phase response genes, but the underlying mechanism for this suppression remains uncertain. Here we report that treatment of human hepatocullular carcinoma HepG2 cells with IL-1beta inhibited the IL-6-dependent binding of
Richard J Calvert et al.
Analytical biochemistry, 343(2), 283-292 (2005-07-16)
Accurate measurement of activity of wild-type K-ras protein is important due to its tumor suppressor action in tissues such as lung. A published method by Taylor and co-workers uses plasmid-containing Escherichia coli cells to produce a glutathione-S-transferase/raf-1 ras binding domain
Shin-Ichiro Miura et al.
Atherosclerosis, 175(2), 235-243 (2004-07-21)
Since we recently demonstrated that high-density lipoprotein induced human coronary artery endothelial cell (HCEC) tube formation through Ras/Raf/ERK (extracellular-signal-regulated kinase) activation [Arterioscler. Thromb. Vasc. Biol. 23 (2003) 802], it is possible that lipid-lowering agents such as statins, which reduce the
Fu-Ming Tsai et al.
Cellular signalling, 18(3), 349-358 (2005-07-12)
The retinoid-inducible gene 1 (RIG1) protein is a retinoid-inducible growth regulator. Previous studies have shown that the RIG1 protein inhibits the signaling pathways of Ras/mitogen-activated protein kinases. However, neither the mode of action nor the site of inhibition of RIG1

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service