Merck
  • Acetate Production from Glucose and Coupling to Mitochondrial Metabolism in Mammals.
Acetate Production from Glucose and Coupling to Mitochondrial Metabolism in Mammals.
Cell (2018-09-25)
Xiaojing Liu, Daniel E Cooper, Ahmad A Cluntun, Marc O Warmoes, Steven Zhao, Michael A Reid, Juan Liu, Peder J Lund, Mariana Lopes, Benjamin A Garcia, Kathryn E Wellen, David G Kirsch, Jason W Locasale
ABSTRACT
Acetate is a major nutrient that supports acetyl-coenzyme A (Ac-CoA) metabolism and thus lipogenesis and protein acetylation. However, its source is unclear. Here, we report that pyruvate, the end product of glycolysis and key node in central carbon metabolism, quantitatively generates acetate in mammals. This phenomenon becomes more pronounced in the context of nutritional excess, such as during hyperactive glucose metabolism. Conversion of pyruvate to acetate occurs through two mechanisms: (1) coupling to reactive oxygen species (ROS) and (2) neomorphic enzyme activity from keto acid dehydrogenases that enable function as pyruvate decarboxylases. Further, we demonstrate that de novo acetate production sustains Ac-CoA pools and cell proliferation in limited metabolic environments, such as during mitochondrial dysfunction or ATP citrate lyase (ACLY) deficiency. By virtue of de novo acetate production being coupled to mitochondrial metabolism, there are numerous possible regulatory mechanisms and links to pathophysiology.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis
Sigma-Aldrich
Pyruvate Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 0.1-1.0 units/mg protein (Lowry)
Sigma-Aldrich
L-Methionine sulfoxide
Sigma-Aldrich
Hydrogen peroxide-18O2 solution, 2-3% in H2O, 90 atom % 18O
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
α-Ketoglutarate Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 0.1-1.0 units/mg protein (Lowry)
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
Aldrithiol-2, 98%
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit