Merck
Phosphatidylserine dynamics in cellular membranes.
Molecular biology of the cell (2012-04-13)
Jason G Kay, Mirkka Koivusalo, Xiaoxiao Ma, Thorsten Wohland, Sergio Grinstein
ABSTRACT
Much has been learned about the role of exofacial phosphatidylserine (PS) in apoptosis and blood clotting using annexin V. However, because annexins are impermeant and unable to bind PS at low calcium concentration, they are unsuitable for intracellular use. Thus little is known about the topology and dynamics of PS in the endomembranes of normal cells. We used two new probes-green fluorescent protein (GFP)-LactC2, a genetically encoded fluorescent PS biosensor, and 1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phospho-L-serine (TopFluor-PS), a synthetic fluorescent PS analogue-to examine PS distribution and dynamics inside live cells. The mobility of PS was assessed by a combination of advanced optical methods, including single-particle tracking and fluorescence correlation spectroscopy. Our results reveal the existence of a sizable fraction of PS with limited mobility, with cortical actin contributing to the confinement of PS in the plasma membrane. We were also able to measure the dynamics of PS in endomembrane organelles. By targeting GFP-LactC2 to the secretory pathway, we detected the presence of PS in the luminal leaflet of the endoplasmic reticulum. Our data provide new insights into properties of PS inside cells and suggest mechanisms to account for the subcellular distribution and function of this phospholipid.
Product Description
Avanti
16:0 Tempo PC, Avanti Research - A Croda Brand 810606P, powder
Avanti
16:0-10 Doxyl PC, Avanti Research - A Croda Brand 810603P, powder
Avanti
C11 TopFluor GM3, Avanti Research - A Croda Brand 810258E, ethanol solution
Avanti
16:0-10 Doxyl PC, Avanti Research - A Croda Brand 810603C
Avanti
TopFluor PS, Avanti Research - A Croda Brand 810283P, powder
Avanti
16:0-5 Doxyl PC, Avanti Research - A Croda Brand 810601C
Avanti
16:0-5 Doxyl PC, Avanti Research - A Croda Brand 810601P, powder
Avanti
16:0-12 Doxyl PC, Avanti Research - A Croda Brand 810600P, powder
Avanti
16:0-7 Doxyl PC, Avanti Research - A Croda Brand 810602P, powder
Avanti
16:0-16 Doxyl PC, Avanti Research - A Croda Brand 810604C
Avanti
TopFluor PC, 1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0-16 Doxyl PC, Avanti Research - A Croda Brand 810604P, powder
Avanti
TopFluor PC, 1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phosphocholine, powder
Avanti
TopFluor PE, Avanti Research - A Croda Brand 810282P, powder
Avanti
TopFluor PE, Avanti Research - A Croda Brand 810282C