Skip to Content
Merck
All Photos(2)

Key Documents

810282C

Avanti

TopFluor PE

Avanti Research - A Croda Brand 810282C

Synonym(s):

1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H77BF2N3O8P
CAS Number:
Molecular Weight:
867.89
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810282C-1mg)
pkg of 5 × 1 mL (810282C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810282C

concentration

1 mg/mL (810282C-1mg)
1 mg/mL (810282C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

TopFluor has a moiety, called dipyrromethene boron difluoride. This fluorescent molecule is covalently attached to various lipid molecules. Phosphoethanolamine is a precursor to phosphatidylcholine and phosphatidylethanolamine.

Application

TopFluor PE may be used as a fluorescently labeled lipid to replace the head labeled lipid (TFPE-head) to study the fusion efficiency of liposomes. It has also been used in the preparation of liposome.

Biochem/physiol Actions

Phosphoethanolamine participates in phospholipid metabolism. It has the ability to repress tumor growth both in vitro and in vivo.

Packaging

5 mL Amber Glass Screw Cap Vial (810282C-1mg)
5 mL Amber Glass Screw Cap Vial (810282C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single Particle Fluorescence Burst Analysis of Epsin Induced Membrane Fission
Brooks A, et al.
PLoS ONE, 10(3) (2015)
Influence of Environmental Conditions on the Fusion of Cationic Liposomes with Living Mammalian Cells
Kolasinac R, et al.
Nanomaterials, 9(7), 1025-1025 (2019)
Phosphoethanolamine and the danger of unproven drugs
Pondee N, et al.
Ecancermedicalscience (2016)
Fumonisin, Folate, and Neural Tube Defects
Comprehensive Toxicology, 12, 189-208 (2010)
Single Particle Fluorescence Burst Analysis of Epsin Induced Membrane Fission
Brooks A, et al.
Testing, 10(3) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service