Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, screw top, clear glass (vial only)
Products
Product Category
vials (14)
Brand
Supelco (14)
Agency
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
854190
Vials, screw top, clear glass (vial only),
volume 4 mL, clear glass vial, thread for 13-425, pkg of 100 ea
Expand
View Pricing
27151
Vials, screw top, clear glass (vial only),
volume 7 mL, clear glass vial, thread for 15-425, pkg of 100 ea
Expand
View Pricing
27078
Vials, screw top, clear glass (vial only),
volume 2 mL, clear glass vial, thread for 8-425, pkg of 100 ea
Expand
View Pricing
27184
Vials, screw top, clear glass (vial only),
volume 40 mL, clear glass vial, thread for 24-400, pkg of 100 ea
Expand
View Pricing
27379
Vials, screw top, clear glass (vial only),
volume 40 mL, clear glass vial, thread for 24-400, pkg of 100 ea
Expand
View Pricing
27173
Vials, screw top, clear glass (vial only),
volume 22 mL, clear glass vial, thread for 20-400, pkg of 100 ea
Expand
View Pricing
27162
Vials, screw top, clear glass (vial only),
volume 15 mL, clear glass vial, thread for 18-400, pkg of 100 ea
Expand
View Pricing
23703-U
Vials, screw top, clear glass (vial only),
volume 60 mL, clear glass vial, thread for 24-400, pkg of 144 ea
Expand
View Pricing
27111
Vials, screw top, clear glass (vial only),
volume 4 mL, clear glass vial, thread for 13-425, O.D. × H × I.D. 15 mm × 45 mm × 8 mm, pkg of 100 ea
Expand
View Pricing
SU860001
Vials, screw top, clear glass (vial only),
volume 40 mL, clear glass vial, thread for 24-400, pkg of 100 ea
Expand
View Pricing
27265
Vials, screw top, clear glass (vial only),
volume 2 mL, clear glass vial, thread for 10-425, O.D. × H × I.D. 12 mm × 32 mm × 6 mm, pkg of 100 ea
Expand
View Pricing
27031
Vials, screw top, clear glass (vial only),
volume 4 mL, clear glass vial, thread for 13-425, O.D. × H × I.D. 15 mm × 45 mm × 8 mm, pkg of 1000 ea
Expand
View Pricing
27079
Vials, screw top, clear glass (vial only),
volume 2 mL, clear glass vial, thread for 8-425, pkg of 1000 ea
Expand
View Pricing
27113
Vials, screw top, clear glass (vial only),
volume 4 mL, clear glass vial (with marking spot), thread for 13-425, pkg of 1000 ea
Expand
View Pricing