Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vial crimp seals, BiMetal (magnetic), 20 mm, with PTFE/silicone septa, pkg 100
Products
Product Category
septa (4)
Brand
Supelco (4)
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
29169-U
Vial crimp seals, BiMetal (magnetic), 20 mm, with PTFE/silicone septa, pkg 100,
red seal, PTFE/silicone septa, diam. × thickness 20 mm × 3 mm
Expand
View Pricing
29176-U
Vial crimp seals, BiMetal (magnetic), 20 mm, with PTFE/silicone septa, pkg 100,
blue seal, PTFE/silicone septa, diam. × thickness 20 mm × 3 mm
Expand
View Pricing
29175-U
Vial crimp seals, BiMetal (magnetic), 20 mm, with PTFE/silicone septa, pkg 100,
gold seal, PTFE/silicone septa, diam. × thickness 20 mm × 3 mm
Expand
View Pricing
29178-U
Vial crimp seals, BiMetal (magnetic), 20 mm, with PTFE/silicone septa, pkg 100,
green seal, PTFE/silicone septa, diam. × thickness 20 mm × 3 mm
Expand
View Pricing