Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tris(2-ethylhexyl) phosphate
Synonym(s):
‘Trioctyl’ phosphate, Tris(2-ethylhexyl) phosphate
Linear Formula:
[CH
3
(CH
2
)
3
CH(C
2
H
5
)CH
2
O]
3
P(O)
CAS Number:
78-42-2
Molecular Weight:
434.63
EC Number:
201-116-6
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
289922
Tris(2-ethylhexyl) phosphate,
97%
Expand
View Pricing
8.21202
Tris(2-ethylhexyl) phosphate,
for synthesis
Expand
View Pricing
93299
Tris(2-ethylhexyl) phosphate,
Selectophore
™
, ≥99.0%
Expand
View Pricing