Skip to Content
Merck
Tetrahydro-4H-pyran-4-one 99%

Tetrahydro-4H-pyran-4-one

Tetrahydro-4H-pyran-4-one 99%
Synonym(s):
4-Oxotetrahydropyran
Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing