
Methyl 6-chloropyridine-3-carboxylate

Synonym(s):
Methyl 6-chloro-3-picolinate, Methyl 6-chloronicotinate
Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing