
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt

Synonym(s):
8-Br-cAMP
Empirical Formula (Hill Notation):
C10H10BrN5NaO6P
CAS Number:
Molecular Weight:
430.08
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing