203800
Adenosine 3′,5′-cyclic Monophosphate, 8-Bromo-, Sodium Salt
Cell-permeable cAMP analog that has greater resistance to phosphodiesterases than cAMP.
Synonym(s):
Adenosine 3′,5′-cyclic Monophosphate, 8-Bromo-, Sodium Salt, 8-Bromo-cAMP, Na
About This Item
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
color
white
solubility
water: soluble
shipped in
ambient
storage temp.
−20°C
InChI
1S/C10H11BrN5O6P.Na/c11-10-15-4-7(12)13-2-14-8(4)16(10)9-5(17)6-3(21-9)1-20-23(18,19)22-6;/h2-3,5-6,9,17H,1H2,(H,18,19)(H2,12,13,14);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1
InChI key
DMRMZQATXPQOTP-GWTDSMLYSA-M
Related Categories
General description
Biochem/physiol Actions
PKA
Warning
Reconstitution
Other Notes
de Rooij, J., et al. 1998. Nature 396, 474.
Boyer, B., and Thiery, J.P. 1993. J. Cell Biol.120, 767.
Hei, Y.-J., et al. 1991. Mol. Pharmacol. 39, 233.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service