1A00070
USP
3′-(N,N-Didemethyl)Azithromycin; (Aminoazithromycin)
Pharmaceutical Analytical Impurity (PAI)
Synonym(s):
((2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-amino-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2- ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)- 3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-13-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3-amino-3,4,6-trideoxy-β-D-xylo-hexopyranosyl]oxy]-1-oxa-6- azacyclopentadecan-15-one)
About This Item
Recommended Products
grade
pharmaceutical analytical impurity (PAI)
Agency
USP
API family
azithromycin
manufacturer/tradename
USP
application(s)
pharmaceutical
format
neat
storage temp.
2-8°C
1 of 4
This Item | 1A00080 | 1A00840 | 1A00210 |
---|---|---|---|
USP | USP | USP | USP |
neat | neat | neat | neat |
pharmaceutical | pharmaceutical | pharmaceutical | pharmaceutical |
USP | USP | USP | USP |
azithromycin | azithromycin | azithromycin | azithromycin |
General description
USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.
Associated Drug Substance: Azithromycin
Therapeutic Area: Antibiotics.
For more information about this PAI, visit here.
Application
Features and Benefits
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service