Skip to Content
Merck
All Photos(1)

Key Documents

1659000

USP

Thimerosal

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thimerosal

manufacturer/tradename

USP

mp

234-237 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thimerosal USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Thimerosal Tincture
  • Thimerosal Topical Aerosol®
  • Thimerosal Topical Solution

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

Aerosol is a registered trademark of Cytec Technology Corp.

Disclaimer

As part of the corporationglobal plan to support the Minamata Convention in eliminatingmercury-containing compounds, as of January 2021 customers will be required toprovide an End-Use Declaration. The sales of thimerosal will be restricted forapproved applications only, such as preservative in vaccine manufacturing,preservative in eye cosmetics, research use within academia (excludingpreservation intent) and as reference material only. The product is notintended for use as a biocide under global biocide regulations, including butnot limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act ,European Biocidal Products Regulation, Canada’s Pest Management RegulatoryAgency, Turkey’s Biocidal Products Regulation,Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR)and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thimerosal Topical Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 4347-4347 (2013)
Thimerosal Topical Aerosol
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 4346-4346 (2018)
Thimerosal
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 4344-4344 (2020)
Thimerosal Tincture
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 4348-4348 (2013)
David A Geier et al.
Biological trace element research, 163(1-2), 28-38 (2014-11-11)
Thimerosal is an organic mercury (Hg)-containing compound (49.55 % Hg by weight) historically added to many multi-dose vials of vaccine as a preservative. A hypothesis testing case-control study evaluated automated medical records in the Vaccine Safety Datalink (VSD) for organic Hg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service