Skip to Content
Merck
All Photos(1)

Key Documents

1612685

USP

Sildenafil Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-[2-Ethoxy-5[(4-methylpiperazin-1-yl)sulfonyl]phenyl]-1-methyl-3-(2-methylpropyl)-6,7-dihydro-1H-pyrazol[4,3-d]pyrimidin-7-one, Isobutyl sildenafil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N6O4S
CAS Number:
Molecular Weight:
488.60
UNSPSC Code:
41116107
NACRES:
NA.24

Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

sildenafil

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C23H32N6O4S/c1-6-33-19-8-7-16(34(31,32)29-11-9-27(4)10-12-29)14-17(19)22-24-20-18(13-15(2)3)26-28(5)21(20)23(30)25-22/h7-8,14-15H,6,9-13H2,1-5H3,(H,24,25,30)

InChI key

MDDMCAQUTIFYAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
RTN350DMN10RTN10
usage

sufficient for 70 purifications

usage

sufficient for 350 purifications

usage

sufficient for 10 purifications

usage

sufficient for 10 purifications

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

-

sustainability

Greener Alternative Product

RTN10

RTN10, RTN70, RTN9602, RTN9604

DMN70

RTN350, RTN70, RTN9602, RTN9604

greener alternative category

Aligned,

greener alternative category

Aligned,

greener alternative category

-

greener alternative category

, Aligned

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sildenafil Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Sildenafil Citrate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service