Skip to Content
Merck
All Photos(1)

Key Documents

1612517

USP

Sertraline hydrochloride racemic mixture

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-Sertraline hydrochloride, 1R,4R)-rel-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17Cl2N · HCl
CAS Number:
Molecular Weight:
342.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sertraline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13

InChI

1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1

InChI key

BLFQGGGGFNSJKA-XHXSRVRCSA-N

Gene Information

human ... SLC6A4(6532)

General description

Sertraline hydrochloride is a SSRI i.e selective serotonin reuptake inhibitor. It is mainly used in treating psychiatric disorders. It also is effective in treating hypotension occurring in disorders like neurocardiogenic syncope and idiopathic orthostatic hypotension which is affected by autonomic dysfunction.

Application

Sertraline hydrochloride racemic mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Sertraline Hydrochloride Tablets
  • Sertraline Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aquatic ecotoxicity of the selective serotonin reuptake inhibitor sertraline hydrochloride in a battery of freshwater test species.
Minagh E
Ecotox. Environ. Safety, 72(2), 434-440 (2009)
Effect of sertraline hydrochloride on dialysis hypotension.
Dheenan S
American Journal of Kidney Diseases, 31(4), 624-630 (1998)
Nachshon Korem et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(12), 2709-2722 (2014-06-06)
Posttraumatic stress disorder (PTSD) can develop following exposure to a traumatic event. Hence, what we do in the first few hours after trauma exposure may alter the trajectory of PTSD. We examined whether cannabinoids can prevent the effects of a
Baolian Wang et al.
Journal of ethnopharmacology, 155(3), 1473-1482 (2014-08-06)
Schisandra chinensis (SC), officially listed as a sedative and tonic in the Chinese Pharmacopoeia, has been used as a common component in various prescriptions in Traditional Chinese Medicine (TCM) and more recently in western medicine for its antihepatotoxic effect. To
Eman G Nouman et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 989, 37-45 (2015-03-24)
Dabigatran etexilate (DABE) is a low-molecular-weight prodrug that is converted after oral administration to dabigatran (DAB)-a directly acting oral anticoagulant. In this study, an LC-MSMS assay was developed and validated for the determination of DABE, free DAB and its equipotent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service