Skip to Content
Merck
All Photos(1)

Key Documents

1604814

USP

Ritonavir Related Compounds Mixture

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Mixture of ritonavir, hydroxyritonavir

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ritonavir

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

mixture

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ritonavir Related Compounds Mixture USP reference standard suitable for use in specified USP compendial tests and assays.

Also used to prepare system suitability stock, system suitability, and identity solution for impurity analysis by using the liquid chromatography method and UV detector according to the given below monographs of United States Pharmacopeia (USP):
  • Lopinavir and Ritonavir Tablets
  • Ritonavir Tablets
  • Ritonavir

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ritonavir Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(4), 3920-3920 (2013)
Ritonavir
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(4), 3912-3912 (2020)
Lopinavir and Ritonavir Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(4), 2670-2670 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service