Skip to Content
Merck

1481215

USP

Oxaliplatin Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Oxalic acid dihydrate, Ethanedioic acid dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
CHF 67.10
10 ML
CHF 484.00

CHF 67.10


Availability

Estimated to ship onFebruary 23, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
CHF 67.10
10 ML
CHF 484.00

About This Item

Linear Formula:
HO2CCO2H · 2H2O
CAS Number:
Molecular Weight:
126.07
Beilstein:
3679436
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CHF 67.10


Availability

Estimated to ship onFebruary 23, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

vapor density

4.4 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

API family

oxaliplatin

form

crystalline

manufacturer/tradename

USP

mp

104-106 °C (lit.)

solubility

water: ~126.1 g/L at 20 °C

application(s)

pharmaceutical (small molecule)

format

neat

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA011408HPA000888HPA008994
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

UniProt accession no.

Q4G0X9

UniProt accession no.

Q96ER9

UniProt accession no.

O60826

UniProt accession no.

-

species reactivity

human

species reactivity

human

species reactivity

human, mouse, rat

species reactivity

human, mouse, rat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxaliplatin Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Oxaliplatin
  • Oxaliplatin for Injection
  • Oxaliplatin Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service