Skip to Content
Merck
All Photos(1)

Key Documents

1478833

USP

Orlistat Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Triphenylphosphine oxide, Ph3PO, TPPO, Triphenyl phosphorus oxide, Triphenylphosphine monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3PO
CAS Number:
Molecular Weight:
278.28
Beilstein:
745854
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

orlistat

manufacturer/tradename

USP

mp

150-157 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15OP/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

FIQMHBFVRAXMOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Orlistat Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Latanoprost
  • Orlistat

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of USP Biologics Program.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service