Skip to Content
Merck
All Photos(1)

Key Documents

1477502

USP

Octoxynol 9

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Polyethylene glycol mono(4-tert-octylphenyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H22O(OCH2CH2)n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

octoxynol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O(CCO)c1ccc(cc1)C(CC(C)(C)C)(C)C

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3

InChI key

JYCQQPHGFMYQCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.Octoxynol 9 is an anhydrous liquid mixture consisting chiefly of mono[p-(1,1,3,3-tetramethylbutyl)] phenyl ethers of polyethylene glycols, corresponding to (CH3)3C(CH2)C(CH3)2C6H4(OCH2CH2)nOH, in which the average value of n is about 9.

Application

It is intended to be used as a USP reference standard in specified quality tests and assays.
Also used to prepare standard and system suitability solutions in the assay of octoxynol 9 as described in the United States Pharmacopeia (USP) monograph.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Octoxynol 9
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(5), 5901-5901 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service