Skip to Content
Merck
All Photos(1)

Key Documents

1444279

USP

Mirtazapine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3
CAS Number:
Molecular Weight:
265.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

mirtazapine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1CCN2C(C1)c3ccccc3Cc4cccnc24

InChI

1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3

InChI key

RONZAEMNMFQXRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mirtazapine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Mirtazapine Tablets
  • Mirtazapine Orally Disintegrating Tablets
  • Mirtazapine Compounded Oral Suspension, Veterinary

Biochem/physiol Actions

Mirtazapine is a noradrenergic and specific serotonergic antidepressant (NaSSA). Mirtazapine agonizes selective adrenergic and serotonergic receptors so that both NE release and 5-HT1A mediated serotonergic signaling are increased.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirtazapine
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(6), 2976-2976 (2021)
Mirtazapine Compounded Oral Suspension, Veterinary
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(5), 2977-2977 (2019)
Mirtazapine Orally Disintegrating Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(2), 2980-2980 (2020)
Katarzyna Kamińska et al.
Pharmacological reports : PR, 66(6), 984-990 (2014-12-03)
The aim of our study was to understand the mechanism of clinical efficacy of the combination of an antidepressant and risperidone in drug-resistant depression. We studied the effect of an antidepressant (mirtazapine) and risperidone (atypical antipsychotic), given separately or jointly
Hun Soo Chang et al.
Journal of clinical psychopharmacology, 34(4), 446-454 (2014-06-10)
We tested for the association of HTR1A and 5-HTT genetic polymorphisms with treatment response to mirtazapine and evaluated the interactive effect between the polymorphisms in 283 patients with major depressive disorder. Korean subjects with diagnosis of major depressive disorder using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service