Skip to Content
Merck
All Photos(1)

Key Documents

1417001

USP

Methoxsalen

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

8-Methoxypsoralen, 8-MOP, 9-Methoxyfuro[3,2-g][1]benzopyran-7-one, Ammoidin, Methoxsalen, Xanthotoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O4
CAS Number:
Molecular Weight:
216.19
Beilstein:
196453
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

methoxsalen

manufacturer/tradename

USP

mp

148-150 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1c2OC(=O)C=Cc2cc3ccoc13

InChI

1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3

InChI key

QXKHYNVANLEOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
56448PHR3040PHL89866
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

PhytoLab

format

neat

format

neat

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

pharmaceutical

application(s)

-

API family

methoxsalen

API family

-

API family

methoxsalen

API family

-

mp

148-150 °C (lit.)

mp

148-150 °C (lit.)

mp

148-150 °C (lit.)

mp

148-150 °C (lit.)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

8-methoxypsoralen (8-MOP) plus ultraviolet A (UVA) irradiation induces monoadducts and interstrand cross-links in DNA and therefore can be used to study DNA repair and recombination mechanisms. Cultured normal human melanocytes treated with 8-methoxypsoralen and irradiated with ultraviolet A (UVA) formed 8-methoxypsoralen-phospholipid photoadducts that could be substituted for diacylglycerol to activate protein kinase C in a cell-free system. 8-MOP is an inactivator of purified reconstituted cytochrome P450. It also inhibits substance P-induced histamine release from substance P-activated rat peritoneal mast cells by suppressing the rise in [Ca2+].

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service