1238002
USP
Equilin
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1,3,5(10),7-Estratetraen-3-ol-17-one, 3-Hydroxy-1,3,5(10),7-estratetraen-17-one, 7-Dehydroestrone
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
equilin
manufacturer/tradename
USP
mp
238-240 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
C[C@]12CC[C@H]3C(=CCc4cc(O)ccc34)[C@@H]1CCC2=O
InChI
1S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3-5,10,14,16,19H,2,6-9H2,1H3/t14-,16+,18+/m1/s1
InChI key
WKRLQDKEXYKHJB-HFTRVMKXSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
1 of 4
This Item | Y0000052 | E8126 | 1260001 |
---|---|---|---|
pharmaceutical primary standard | pharmaceutical primary standard | - | pharmaceutical primary standard |
neat | neat | - | neat |
pharmaceutical (small molecule) | pharmaceutical (small molecule) | - | pharmaceutical (small molecule) |
equilin | equilin | - | ethinyl estradiol |
238-240 °C (lit.) | 238-240 °C (lit.) | 238-240 °C (lit.) | 182-183 °C (lit.) |
General description
Application
- Conjugated Estrogens
- Conjugated Estrogens Tablets
- Equilin
- Esterified Estrogens
- Esterified Estrogens Tablets
Analysis Note
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Lact. - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service