Skip to Content
Merck
All Photos(1)

Key Documents

1075677

USP

Bisoctrizole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,2′-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H50N6O2
CAS Number:
Molecular Weight:
658.87
Beilstein:
11326835
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

bisoctrizole

manufacturer/tradename

USP

mp

197-199 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(C)CC(C)(C)c1cc(Cc2cc(cc(c2O)-n3nc4ccccc4n3)C(C)(C)CC(C)(C)C)c(O)c(c1)-n5nc6ccccc6n5

InChI

1S/C41H50N6O2/c1-38(2,3)24-40(7,8)28-20-26(36(48)34(22-28)46-42-30-15-11-12-16-31(30)43-46)19-27-21-29(41(9,10)25-39(4,5)6)23-35(37(27)49)47-44-32-17-13-14-18-33(32)45-47/h11-18,20-23,48-49H,19,24-25H2,1-10H3

InChI key

FQUNFJULCYSSOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bisoctrizole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service