All Photos(1)
About This Item
Linear Formula:
(NaPO3)n
Molecular Weight:
617.82
UNSPSC Code:
12352302
grade:
reagent grade
Recommended Products
grade
reagent grade
product line
uniLAB™
packaging
poly bottle of 1 × 500 g (SAAR5822780EM)
storage temp.
2-30°C
Application
Chemically pure
For laboratory use only
Legal Information
uniLAB is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service