Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5821010

Sodium acetate trihydrate

univAR

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
12352302

product line

univAR

autoignition temp.

1112 °F

packaging

poly bottle of 12 × 500 g (SAAR5821010EM)

storage temp.

2-30°C

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service