Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5075040

Propan-2-ol

univAR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHOHCH3
Molecular Weight:
60.10
UNSPSC Code:
12191601

product line

univAR

packaging

glass bottle of 4 × 2.5 L (SAAR5075040LC)
pkg of 25 L (SAAR5075040ME)

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service