Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5044420

Potassium hydroxide

univAR

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352320

vapor pressure

1 mmHg ( 719 °C)

product line

univAR

packaging

poly bottle of 12 × 500 g (SAAR5044420EM)

mp

361 °C (lit.)

storage temp.

2-30°C

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service