Skip to Content
Merck
All Photos(1)

Key Documents

SAAR4124000

Magnesium sulfate heptahydrate

univAR

Synonym(s):

Magnesium sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO4S · 7H2O
Molecular Weight:
246.47
UNSPSC Code:
12352302

product line

univAR

packaging

poly bottle of 12 × 500 g (SAAR4124000EM)

storage temp.

2-30°C

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service