All Photos(1)
About This Item
Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
MDL number:
UNSPSC Code:
12352303
Recommended Products
product line
univAR™
packaging
poly bottle of 1 × 250 g (SAAR4123520DN)
mp
2852 °C (lit.)
storage temp.
2-30°C
SMILES string
O=[Mg]
InChI
1S/Mg.O
InChI key
CPLXHLVBOLITMK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Analysed reagent
For laboratory use only
Legal Information
univAR is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service