Skip to Content
Merck
All Photos(1)

Key Documents

SAAR1021020

Acetic acid, glacial

univAR

Synonym(s):

Acetic acid, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352106

vapor density

2.07 (vs air)

product line

univAR

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

packaging

glass bottle of 1 × 2.5 L (SAAR1021020LC)

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service