PHR9208
4-[4-(4-Chlorophenyl)-3,6-dihydropyridin-1(2H)-yl]-N,N-dimethyl-2,2-diphenylbutanamide
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Loperamide Hydrochloride impurity H (PhEur), 4-(4-Chlorophenyl)-3,6-dihydro-N,N-dimethyl-α,α-diphenyl-1(2H)-pyridinebutanamide
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
pharmaceutical small molecule
storage temp.
-10 to -25°C
SMILES string
O=C(N(C)C)C(C1=CC=CC=C1)(C2=CC=CC=C2)CCN3CC=C(C4=CC=C(Cl)C=C4)CC3
InChI
1S/C29H31ClN2O/c1-31(2)28(33)29(25-9-5-3-6-10-25,26-11-7-4-8-12-26)19-22-32-20-17-24(18-21-32)23-13-15-27(30)16-14-23/h3-17H,18-22H2,1-2H3
InChI key
UUHAGHSPKICYQY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service