PHL84258
Sibiricose A6
phyproof® Reference Substance
Synonym(s):
3-O-Sinapoylsucrose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C23H32O15
CAS Number:
Molecular Weight:
548.49
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
form
solid
manufacturer/tradename
PhytoLab
SMILES string
O1[C@]([C@H]([C@@H]([C@H]1CO)O)OC(=O)\C=C\c3cc(c(c(c3)OC)O)OC)(O[C@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)O)O)CO)CO
InChI key
WTCVROXOIQEIRC-IBVGEFGBSA-N
Related Categories
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service