All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
−20°C
SMILES string
O(C)c1c(ccc(c1)CCC(=O)\C=C\CCCCCCC)O
InChI
1S/C19H28O3/c1-3-4-5-6-7-8-9-10-17(20)13-11-16-12-14-18(21)19(15-16)22-2/h9-10,12,14-15,21H,3-8,11,13H2,1-2H3/b10-9+
InChI key
LGZSMXJRMTYABD-MDZDMXLPSA-N
Related Categories
General description
This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Other Notes
This compound is commonly found in plants of the genus: zingiber
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service