Skip to Content
Merck
All Photos(1)

Key Documents

MSCAL6

Supelco

ProteoMass LTQ/FT-Hybrid ESI Neg. Mode Cal Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

product line

ProteoMass

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 1 × 10 mL

storage condition

protect from light

concentration

(varied conc.)

technique(s)

mass spectrometry (MS): suitable

format

multi-component solution

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The ProteoMass ESI Calibration Kit for LTQ based Hybrids, Negative Ion Mode Calibration Solution, enables calibration of Thermo Scientific hybrid instruments equipped with ESI source in negative ion mode. It covers a mass range of m/z 265 to m/z 1880, and is therefore usable for calibrations between m/z 50 and 2000.

Features and Benefits

Discover the Advantages for Yourself!
  • Ready-to-use
  • Developed and qualified specifically for use with the LTQ based Thermo Scientific hybrid instruments (LTQ FT®, LTQ FT Ultra®, and LTQ Orbitrap series) with ESI source and Thermo Scientific Exactive
  • Conveniently packaged, freeing you from time-consuming mixing and dilution steps, allowing you to focus on acquisition of quality data

Legal Information

LTQ FT is a registered trademark of Thermo Fisher Scientific or its subsidiaries
LTQ FT ULTRA is a registered trademark of Thermo Fisher Scientific or its subsidiaries
LTQ Orbitrap is a trademark of Thermo Fisher Scientific or its subsidiaries
ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.1 °F - closed cup

Flash Point(C)

5.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Cécile Alexandre-Gouabau et al.
Nutrients, 10(2) (2018-02-01)
Human milk is recommended for feeding preterm infants. The current pilot study aims to determine whether breast-milk lipidome had any impact on the early growth-pattern of preterm infants fed their own mother's milk. A prospective-monocentric-observational birth-cohort was established, enrolling 138
Deok-Kyu Hwang et al.
Pharmaceutics, 11(7) (2019-07-25)
Catalposide, an active component of Veronica species such as Catalpa ovata and Pseudolysimachion lingifolium, exhibits anti-inflammatory, antinociceptic, anti-oxidant, hepatoprotective, and cytostatic activities. We characterized the in vitro metabolic pathways of catalposide to predict its pharmacokinetics. Catalposide was metabolized to catalposide
Luca Narduzzi et al.
Metabolites, 9(12) (2019-12-01)
Hydrophilic Interaction Liquid Chromatography (HILIC) chromatography is widely applied in metabolomics as a complementary strategy to reverse phase chromatography. Nevertheless, it still faces several issues in terms of peak shape and compounds ionization, limiting the automatic de-convolution and data semi-quantification
Luca Narduzzi et al.
Drug testing and analysis, 12(7), 887-899 (2020-04-05)
Growth hormone (GH), an endogenous peptide regulating anabolism and lipolysis in humans, is known to be abused by athletes to improve their performance. Despite the development of two distinct screening methods, few positive cases have been reported by the antidoping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service