ERMBF411G
Maize GMO Standard ERM-BF411
ERM®, certified reference material, 100% Bt-176, for the detection of GMO maize (Br-176) with PCR
Synonym(s):
GMO Standard ERM-BF411, Maize Bt-176
About This Item
Recommended Products
grade
certified reference material
Quality Level
Agency
ERM®
quality
for the detection of GMO maize (Br-176) with PCR
manufacturer/tradename
JRC
application(s)
food and beverages
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
ERMBF411G
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service