Skip to Content
Merck
All Photos(1)

Key Documents

CRM47792

Supelco

Linolenic Acid Methyl Ester Isomer Mix

certified reference material, 10 mg/mL in dichloromethane, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

Assay

100%

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

ester

shipped in

wet ice

storage temp.

-10 to -25°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Check out our complete portfolio of Fatty acid methyl ester (FAME) reference material solution mixes

Application

The CRM has been used as follows:

  • Study of fatty acid profile of 69 different cold-pressed vegetable oil samples by gas chromatography coupled with flame ionization detection
  • Development and validation of a GC-MS method to simultaneously determine C18 fatty acids in 20 raw milk and 30 commercial milk samples
  • Identification of fatty acid methyl ester peaks in the GC-MS analysis of C18 fatty acids in milk samples to evaluate the hydrogenation of α-Linolenic acid in the rumen of dairy cows
  • Fatty acid analysis of 320 goat, cow, and sheep cheese samples by gas chromatography-flame ionization detection as per the protocol described in AOAC Official Method 2012.13
  • GC-FID analysis of typical Indian ready-made snacks to determine their fatty acid composition based on Association of Official Analytical Chemists (AOAC) Official Method 996.06

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service