Skip to Content
Merck
All Photos(1)

Key Documents

CRM01303

Supelco

Cyantraniliprole

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3-Bromo-1-(3-chloro-2-pyridyl)-4′-cyano-2′-methyl-6′-(methylcarbamoyl)pyrazole-5-carboxanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14BrClN6O2
CAS Number:
Molecular Weight:
473.71
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

O=C(C1=CC(Br)=NN1C2=C(Cl)C=CC=N2)NC3=C(C(NC)=O)C=C(C#N)C=C3C

InChI

1S/C19H14BrClN6O2/c1-10-6-11(9-22)7-12(18(28)23-2)16(10)25-19(29)14-8-15(20)26-27(14)17-13(21)4-3-5-24-17/h3-8H,1-2H3,(H,23,28)(H,25,29)

InChI key

DVBUIBGJRQBEDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. The CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Cyantraniliprole, a second-generation diamide insecticide, activates the ryanodine receptors of insects to cause the release of Ca2+ ions resulting in muscle paralysis and death of the insect. It shows ovicidal, larvicidal, and adulticide activity, depending upon the pest species. It is used for cereal crops and vegetables, such as rice, peanuts, corn, tomatoes, green onions, and cucumbers for protection against lepidopteran and coleopteran pests.

Cyantraniliprole is approved for use in the European Union. The maximum residue limits (MRL) for cyantraniliprole in food crops, as per EU regulations are in the range of 0.01-15 mg/kg. As per a request received for cyantraniliprole MRL modification by an evaluating member state (EMS) on 28th January 2015, the EFSA has proposed an MRL of 0.5 mg/kg in strawberries, 0.2 mg/kg in the crop groups “herbal infusions from roots” and “root and rhizome spices”, 0.04 mg/kg on beans without pods, 0.15 mg/kg on peas without pods, and 0.1 mg/kg in globe artichokes. No MRL modifications were made for cherries, carrots, root and tuber vegetables, and brussels sprouts. Cyanantraniliprole is monitored in the Multiannual Control Programmes for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques.
Cyantraniliprole certified reference material (CRM) may also find following use:
  • Study of DNA damage, oxidative stress, and biological toxicity caused in earthworm by cyantraniliprole
  • Development of an isotope-labelled internal standard based method using ultra-high-performance liquid chromatography-tandem mass spectrometry for the analysis of chlorantraniliprole and cyantraniliprole in fruits, vegetables, and cereals
  • Simultaneous determination of 8 new generation amide insecticides by ultrahigh performance liquid chromatography-tandem mass spectrometry in complex food matrices
  • Estimation of five diamide insecticides using carbon nanotube multiplug filter for sample clean-up and ultrahigh-performance liquid chromatography-tandem mass spectrometry in food matrices
  • Simultaneous analysis of five diamide insecticides in edible mushrooms by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) and a modified QuEChERS procedure for extraction

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhou Lu et al.
Journal of agricultural and food chemistry, 67(39), 10977-10983 (2019-09-07)
In this study, an analytical method was developed and validated for simultaneous determination of five diamide insecticides (chlorantraniliprole, cyantraniliprole, flubendiamide, cyclaniliprole, and tetrachlorantraniliprole) in food matrices. Determination of the latter two diamide compounds is first reported. Samples were cleaned up
Modification of the existing maximum residue levels for cyantraniliprole in various crops
European Food Safety Authority (EFSA)
EFSA Journal, 4263 null
Xinglu Pan et al.
Analytical and bioanalytical chemistry, 407(14), 4111-4120 (2015-03-31)
A reliable and sensitive isotope-labelled internal standard method for simultaneous determination of chlorantraniliprole and cyantraniliprole in fruits (apple and grape), vegetables (cucumber and tomato) and cereals (rice and wheat) using ultra-high-performance liquid chromatography-tandem mass spectrometry was developed. Isotope-labelled internal standards
Fajun Tian et al.
Food chemistry, 333, 127468-127468 (2020-07-14)
In this study, a new method for simultaneous determination of cyantraniliprole, chlorantraniliprole, tetrachlorantraniliprole, cyclaniliprole and flubendiamide in edible mushrooms by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) combined with a modified QuEChERS procedure. The samples were extracted using
Tao Lin et al.
Scientific reports, 11(1), 23208-23208 (2021-12-03)
Eight new generation amide insecticide residues analysis by multiwalled carbon nanotubes (MWCNs) cleanup, combined with QuEChERS and ultrahigh performance liquid chromatography tandem mass spectrometry has been developed and successfully applied in complex matrix such as orange, celery, onion, litchi, mango

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service