LpDNPH S10 Cartridge
S10
Synonym(s):
LpDNPH S10 Cartridge Tube
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41104925
Recommended Products
packaging
pkg of 1 × 10 ea (21026-U)
application(s)
environmental
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service