Skip to Content
Merck
All Photos(1)

Key Documents

95711

Supelco

1,3,5-Trioxane

analytical standard

Synonym(s):

Metaformaldehyde, Trioxymethylene, sym.-Trioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
102769
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

777 °F

expl. lim.

29 %

technique(s)

HPLC: suitable

mp

59-62 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

C1OCOCO1

InChI

1S/C3H6O3/c1-4-2-6-3-5-1/h1-3H2

InChI key

BGJSXRVXTHVRSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Repr. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service