Skip to Content
Merck
All Photos(1)

Key Documents

819665

TSKgel® SuperHZM-H (10 µm) HPLC Columns

L x I.D. 15 cm × 6 mm, polystyrene-divinylbenzene phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® SuperHZM-H HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 15 cm × 6 mm, 10 μm particle size, stainless steel hardware

material

stainless steel hardware

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 19665

parameter

0.7 mL/min flow rate
10 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

15 cm × 6 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

10 μm

pore size

mixed

separation technique

size exclusion (SEC)

General description

TSKgel SuperHZM-H mixed-bed columns are packed with rigid, porous, polymer beads each having a varying mean pore diameter. They are used for the non-aqueous separation of organic-soluble polymers with high molecular weights of less than 400,000,000 Da. The THF shipping solvent may be replaced by a limited number of organic solvents.

Physical form

THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane.

Other Notes

Uses guard 819667, available as a custom product. Please inquire.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service