Skip to Content
Merck
All Photos(1)

Key Documents

78991

Supelco

Silica gel Inorganic Sorbent

spherical, 70-200 mesh

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

product name

Silica gel spherical, 75-200 μm particle size

grade

high-purity grade

Quality Level

form

powder

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

surface area

250-350 m2/g

loss

~5% loss on drying

matrix

Silica

matrix active group

silica

particle size

75-200 μm

pore size

0.7-0.9 cm3/g pore volume
110 Å pore size

pH

~7 (5% in H2O, suspension)

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

High purity grade silica gel offers the lowest moisture content, tightest particle size distribution, minimal impurities and greatest lot-to-lot consistency which results in greater reproducibility of the separation process. It is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service