Skip to Content
Merck
All Photos(1)

Key Documents

72971

Supelco

Oxydemeton-methyl Metabolite

PESTANAL®, analytical standard

Synonym(s):

Demeton-​S-​methyl sulfoxide, O,O-Dimethyl S-[2-(ethylsulfinyl)ethyl]monothiophosphate, O,O-Dimethyl S-[2-(ethylsulfinyl)ethyl]monothiophosphate, Demeton-S-methyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15O4PS2
CAS Number:
Molecular Weight:
246.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0 % (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

For R&D, and Analytical use only. Not for drug, household or other uses

Application

Metabolite for food testing

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service