Skip to Content
Merck
All Photos(1)

Key Documents

58220-U

Supelco

SUPELCOSIL LC-8 (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Synonym(s):

LC-8 Reverse-Phase HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

SUPELCOSIL LC-8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L7

product line

SUPELCOSIL

feature

endcapped

packaging

1 ea of

extent of labeling

6.0% carbon loading

parameter

≤70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.2 μmol/m2

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

A phase less hydrophobic than C18. Provides less retention of both polar and non-polar compounds than C18. Use a mobile phase containing 5% less organic modifier for the C8 column than C18. Polar compounds are, relatively, more strongly retained on C8 than C18 columns.

Application


  • Chromatographic behaviour of ionic liquid cations in view of quantitative structure-retention relationship.: This study explores the chromatographic behavior of ionic liquid cations using quantitative structure-retention relationships (QSRR). The SUPELCOSIL LC-8 HPLC Column was utilized to analyze retention data for various ionic liquids, providing insights into their separation mechanisms and potential applications in analytical chemistry (Molíková et al., 2010).

  • Liquid chromatographic method for the determination of lidocaine and monoethylglycine xylidide in human serum containing various concentrations of bilirubin for the assessment of liver function.: The research presents a liquid chromatographic method using the SUPELCOSIL LC-8 HPLC Column to determine lidocaine and its metabolite in human serum. This method is particularly useful for assessing liver function by measuring drug metabolism (Piwowarska et al., 2004).

  • LC determination of morphine and morphine glucuronides in human plasma by coulometric and UV detection.: The research describes a method for the determination of morphine and its glucuronides in human plasma. Utilizing the SUPELCOSIL LC-8 HPLC Column, the study demonstrates the method′s sensitivity and applicability in pharmaceutical analysis (Ary et al., 2001).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thomas Borch et al.
Journal of chromatography. A, 1022(1-2), 83-94 (2004-02-03)
Explosives such as 2,4,6-trinitrotoluene (TNT), octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX), and hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) are widely distributed environmental contaminants. Complete chromatographic separation is necessary in order to accurately determine and quantify explosives and their degradation products in environmental samples and in (bio)transformation studies. The
F Belal et al.
Journal of pharmaceutical and biomedical analysis, 24(3), 335-342 (2001-02-24)
A simple, rapid and sensitive HPLC method has been developed for the simultaneous determination of ramipril and hydrochlorothiazide in their dosage forms. Acetonitrile: sodium perchlorate solution (0.1 M) adjusted to pH 2.5+/-0.2 with phosphoric acid (46:54 v/v), was used as
Jadwiga Piwowarska et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 1-5 (2004-04-29)
A high-performance liquid chromatographic method is described for determination of lidocaine (2-(dietyloamino)-N-(2,6-dimetylofenylo) acetamid) and its metabolite, monoethylglycine xylidide (MEGX), in human serum containing various concentration of bilirubin. Lidocaine and its metabolite were extracted from human serum using dichloromethane. After separation
K Ary et al.
Journal of pharmaceutical and biomedical analysis, 26(2), 179-187 (2001-07-27)
A reversed-phase high-performance liquid chromatographic method with coulometric and UV detection has been developed for the simultaneous determination of morphine, morphine-3-glucuronide and morphine-6-glucuronide. The separation was carried out by using a Supelcosil LC-8 DB reversed-phase column and 0.1 M potassium
P K Kunicki
Journal of chromatography. B, Biomedical sciences and applications, 750(1), 41-49 (2001-02-24)
A HPLC-UV determination of clobazam and N-desmethylclobazam in human serum and urine is presented. After simple liquid-liquid extraction with dichloromethane the compounds and an internal standard diazepam were separated on a Supelcosil LC-8-DB column at ambient temperature under isocratic conditions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service